Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016780.2 | NP_058060.2 | 3 | 310 | ||
GenBank mRNA | AF026509.1 | - | 3 | 254 | ||
AK157958.1 | - | 3 | 256 | |||
AY541673.1 | - | 3 | 224 | |||
BC125518.1 | - | 3 | 310 | 40130033 | ||
BC125520.1 | - | 3 | 310 | 40130035 | ||
U83167.1 | - | 3 | 221 |