Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Park2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016694.3 | NP_057903.1 | 3 | 331 | ||
GenBank mRNA | AB019558.1 | - | 3 | 302 | ||
AF250293.1 | - | 3 | 331 | |||
AF250294.1 | - | 3 | 331 | |||
AF250295.1 | - | 3 | 331 | |||
BC113204.1 | - | 3 | 314 | 40047891 | ||
FJ555554.1 | - | 1 | 81 | |||
FJ555557.1 | - | 3 | 302 |