Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhoa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016802.4 | NP_058082.2 | 3, 4 | 590 | ||
GenBank mRNA | AF014371.1 | - | 2, 3 | 298 | ||
AF178958.1 | - | 2, 3 | 185 | |||
AF178959.1 | - | 2, 3 | 185 | |||
AF178960.1 | - | 2, 3 | 185 | |||
AF178961.1 | - | 2, 3 | 185 | |||
AK077606.1 | - | 3, 4 | 398 | |||
AK083624.1 | - | 3, 4 | 397 | |||
AK150294.1 | - | 3, 4 | 435 | |||
AK150475.1 | - | 3, 4 | 441 | |||
AK152629.1 | - | 3, 4 | 377 | |||
AK165512.1 | - | 3, 4 | 373 | |||
AK172051.1 | - | 3, 4 | 398 | |||
BC068115.1 | - | 3, 4 | 590 | 6809149 | ||
BC096423.1 | - | 3, 4 | 376 | 3590185 | ||
JN971019.1 | - | 1, 2 | 164 |