Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016879.2 | NP_058575.2 | 2, 3 | 700 | ||
GenBank mRNA | AF021836.1 | - | 2, 3 | 684 | ||
AK009099.1 | - | 2, 3 | 699 | |||
AK028825.1 | - | 2, 3 | 700 |