Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016908.2 | NP_058604.1 | 3 | 570 | ||
GenBank mRNA | AB026806.1 | - | 3 | 314 | ||
BC047148.1 | - | 3 | 570 | 6465481 |