Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001150749.1 | NP_001144221.1 | 3 | 778 | ||
NM_017473.4 | NP_059501.1 | 3 | 750 | |||
GenBank mRNA | AB015165.1 | - | 3 | 866 | ||
AB032058.1 | - | 3 | 658 | |||
AF056194.1 | - | 3 | 755 | |||
BC024603.1 | - | 3 | 728 | 4196075 | ||
BC092255.1 | - | 3 | 704 | 5100891 | ||
BC093514.1 | - | 3 | 692 | 5132290 |