Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018743.4 | NP_061213.2 | 13 | 3596 | ||
GenBank mRNA | AK045235.1 | - | 13 | 3490 | ||
AK083589.1 | - | 13 | 3491 |