Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253381.1 | NP_001240310.1 | 6 | 1346 | ||
NM_017377.5 | NP_059073.1 | 6 | 1311 | |||
GenBank mRNA | AB019541.2 | - | 4 | 850 | ||
AF142670.1 | - | 5 | 955 | |||
AK145250.1 | - | 7 | 1368 | |||
AK165829.1 | - | 6 | 1305 | |||
BC138567.1 | - | 5 | 947 | 8861587 | ||
BC138569.1 | - | 5 | 947 | 8861589 | ||
BC145335.1 | - | 5 | 982 | 9053891 |