Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113486.1 | NP_001106958.1 | 12 | 2637 | ||
NM_001113487.1 | NP_001106959.1 | 11 | 2817 | |||
NM_001113488.1 | NP_001106960.1 | 10 | 1986 | |||
NM_017380.2 | NP_059076.1 | 10 | 1967 | |||
GenBank mRNA | AJ250723.1 | - | 10 | 1973 | ||
AK031757.1 | - | 10 | 1862 | |||
AK122415.2 | - | 11 | 2611 | |||
AK141312.1 | - | 12 | 2578 | |||
AK164581.1 | - | 9 | 4581 | |||
BC046524.1 | - | 12 | 2637 | 5290621 | ||
BC094455.1 | - | 13 | 2341 |