Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017461.2 | NP_059489.2 | 7 | 626 | ||
GenBank mRNA | BB610735.1 | - | 7 | 626 | ||
BC140998.1 | - | 7 | 627 | 9055610 | ||
BC145081.1 | - | 7 | 627 | 9053629 | ||
M37030.1 | - | 7 | 630 |