Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001150749.1 | NP_001144221.1 | 3 | 833 | ||
NM_017473.4 | NP_059501.1 | 3 | 805 | |||
GenBank mRNA | AB015165.1 | - | 3 | 921 | ||
AB032058.1 | - | 3 | 713 | |||
AF056194.1 | - | 3 | 810 | |||
BC024603.1 | - | 3 | 783 | 4196075 | ||
BC092255.1 | - | 3 | 759 | 5100891 | ||
BC093514.1 | - | 3 | 747 | 5132290 |