Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018757.1 | NP_061227.1 | 2, 3 | 219 | ||
GenBank mRNA | AF051942.1 | - | 2, 3 | 219 | ||
BC002007.1 | - | 2, 3 | 219 | 3593804 |