Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018773.2 | NP_061243.1 | 2 | 282 | ||
GenBank mRNA | AB014485.1 | - | 2 | 199 | ||
AF051324.1 | - | 2 | 197 | |||
AK076000.1 | - | 2 | 227 | |||
AK144289.1 | - | 2 | 219 | |||
AK172297.1 | - | 2 | 266 | |||
BC003711.1 | - | 2 | 282 | 3599914 |