Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005038.2 | NP_005029.1 | 2 | 289 | ||
GenBank mRNA | AK313929.1 | - | 2 | 290 | ||
BC014893.1 | - | 2 | 276 | |||
BC030707.2 | - | 2 | 285 | 4819748 | ||
BG699755.1 | - | 2 | 294 | |||
CR542081.1 | - | 2 | 177 | |||
L11667.1 | - | 2 | 276 |