Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018864.5 | NP_061352.2 | 9 | 1713 | ||
GenBank mRNA | AF042730.1 | - | 9 | 1658 | ||
AK011770.1 | - | 9 | 1651 | |||
AK078406.1 | - | 9 | 1661 | |||
AK083238.1 | - | 9 | 1648 | |||
AK145472.1 | - | 1651 | ||||
AK147778.1 | - | 9 | 1673 | |||
AK152774.1 | - | 9 | 1664 | |||
AK153457.1 | - | 9 | 1680 | |||
AK160203.1 | - | 9 | 1651 | |||
AK160467.1 | - | 9 | 1646 | |||
AK165977.1 | - | 9 | 1654 | |||
AK168457.1 | - | 9 | 1688 | |||
BC049080.1 | - | 8 | 2455 | 6419835 | ||
BC055722.1 | - | 8 | 1608 | 6413389 |