Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019424.2 | NP_062297.1 | 5 | 529 | ||
GenBank mRNA | AK031397.1 | - | 5 | 519 | ||
AK053974.1 | - | 3 | 361 | |||
AK154994.1 | - | 5 | 694 | |||
AK171225.1 | - | 5 | 508 | |||
BC046329.1 | - | 3 | 353 | |||
U78315.1 | - | 4 | 467 | |||
U97149.1 | - | 4 | 478 |