Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc93b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161428.1 | NP_001154900.1 | 3 | 367 | ||
NM_019449.2 | NP_062322.2 | 3 | 367 | |||
GenBank mRNA | AK030488.1 | - | 3 | 328 | ||
AK039526.1 | - | 3 | 328 | |||
AK155556.1 | - | 3 | 325 | |||
AK162535.1 | - | 3 | 320 | |||
AK165060.1 | - | 3 | 327 | |||
BC018388.1 | - | 3 | 334 | 4209296 |