Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dll4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019454.3 | NP_062327.2 | 4 | 774 | ||
GenBank mRNA | AB043893.1 | - | 4 | 417 | ||
AF253469.1 | - | 4 | 729 | |||
AF273454.1 | - | 4 | 795 | |||
AK004739.1 | - | 4 | 742 | |||
AK031699.1 | - | 4 | 743 | |||
AK052322.1 | - | 4 | 775 | |||
AK165517.1 | - | 4 | 743 | |||
AK171717.1 | - | 4 | 743 | |||
BC042497.1 | - | 4 | 729 | 4017786 | ||
BC049130.1 | - | 4 | 734 | 6825525 |