Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uchl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159866.1 | NP_001153338.1 | 3 | 341 | ||
NM_019562.2 | NP_062508.2 | 3 | 341 | |||
GenBank mRNA | AF148447.1 | - | 3 | 284 | ||
AF175903.1 | - | 3 | 293 | |||
AK007860.1 | - | 2 | 155 | |||
AK011117.1 | - | 3 | 308 | |||
AK017925.1 | - | 3 | 339 | |||
BC006891.1 | - | 3 | 294 | 2650799 | ||
BY279731.1 | - | 3 | 341 |