Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pqbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252528.1 | NP_001239457.1 | 3 | 224 | ||
NM_001252529.1 | NP_001239458.1 | 3 | 290 | |||
NM_019478.4 | NP_062351.2 | 3 | 419 | |||
GenBank mRNA | AJ250406.1 | - | 2 | 170 | ||
AK077652.1 | - | 3 | 249 | |||
BC009657.1 | - | 2 | 192 | 3484619 | ||
BC051673.1 | - | 3 | 229 | 30015428 | ||
BC157976.1 | - | 3 | 324 | 9088137 | ||
BQ550428.1 | - | 3 | 295 | |||
CJ143105.1 | - | 3 | 225 | |||
CN718083.1 | - | 2 | 204 |