Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabggta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019519.2 | NP_062392.1 | 9 | 1325 | ||
GenBank mRNA | AF127656.1 | - | 10 | 1173 | ||
AF127657.1 | - | 9 | 1116 | |||
AF127658.1 | - | 9 | 1325 | |||
AF127659.1 | - | 9 | 1325 | |||
AF127660.1 | - | 10 | 1173 | |||
AF127661.1 | - | 9 | 1116 | |||
AF127662.1 | - | 10 | 1244 | |||
AK002625.1 | - | 10 | 1107 | |||
AK146917.1 | - | 10 | 1196 | |||
BC012214.1 | - | 9 | 1262 | 3967218 |