Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actl6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019673.2 | NP_062647.2 | 2, 3 | 408 | ||
GenBank mRNA | AF041476.1 | - | 2, 3 | 228 | ||
AK002724.1 | - | 2, 3 | 204 | |||
AK090154.1 | - | 2, 3 | 227 | |||
AK167905.1 | - | 2, 3 | 222 | |||
AK169278.1 | - | 2, 3 | 227 | |||
AK182047.1 | - | 1, 2 | 78 | |||
AK205745.1 | - | 1, 2 | 66 | |||
BC001994.1 | - | 2, 3 | 196 | 3491205 | ||
BC094498.1 | - | 2, 3 | 224 | 4209491 | ||
CF725603.1 | - | 2, 3 | 408 |