Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019737.2 | NP_062711.1 | 2 | 1093 | ||
GenBank mRNA | AF097158.1 | - | 2 | 1091 | ||
AF142674.1 | - | 2 | 301 | |||
AK137385.1 | - | 2 | 1073 | |||
AK138247.1 | - | 2 | 991 | |||
AK138610.1 | - | 2 | 1075 | |||
AK144997.1 | - | 2 | 967 | |||
AK163956.1 | - | 2 | 1032 | |||
AK164262.1 | - | 2 | 1032 | |||
AK164724.1 | - | 2 | 1032 | |||
BC011149.1 | - | 2 | 316 | 4037384 |