Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019741.3 | NP_062715.2 | 5 | 835 | ||
GenBank mRNA | AF161071.1 | - | 3 | 196 | ||
AK029720.1 | - | 5 | 836 | |||
BC023500.1 | - | 3 | 211 | 5043284 |