Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001935.2 | NP_001001935.1 | 4 | 581 | ||
NM_001001937.1 | NP_001001937.1 | 5 | 531 | |||
NM_001257334.1 | NP_001244263.1 | 4 | 508 | |||
NM_001257335.1 | NP_001244264.1 | 4 | 844 | |||
NM_004046.5 | NP_004037.1 | 4 | 508 | |||
XM_017025789.1 | XP_016881278.1 | 4 | 450 | |||
GenBank mRNA | AK092735.1 | - | 4 | 523 | ||
AK098765.1 | - | 4 | 440 | |||
AK129739.1 | - | 4 | 451 | |||
AK129900.1 | - | 4 | 451 | |||
AK289457.1 | - | 4 | 777 | |||
AK294804.1 | - | 2 | 594 | |||
AK302272.1 | - | 4 | 451 | |||
AK310051.1 | - | 4 | 806 | |||
AU120644.1 | - | 4 | 476 | |||
AU143001.1 | - | 5 | 525 | |||
BC008028.2 | - | 1 | 78 | |||
BC011384.1 | - | 4 | 439 | 3867375 | ||
BC016046.1 | - | 4 | 426 | 4646761 | ||
BC019310.1 | - | 4 | 439 | 2823663 | ||
BC028335.1 | - | 4 | 428 | |||
BC039135.1 | - | 4 | 421 | 3873390 | ||
BC042694.1 | - | 4 | 421 | |||
BC064562.1 | - | 4 | 439 | 5582043 | ||
BC067385.1 | - | 4 | 451 | 5491013 | ||
BT007209.1 | - | 4 | 386 | |||
CD244623.1 | - | 4 | 467 | |||
D14710.1 | - | 4 | 449 | |||
FJ224309.1 | - | 4 | 444 | |||
X59066.1 | - | 4 | 444 | |||
X65460.1 | - | 4 | 400 |