Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019718.2 | NP_062692.1 | 3 | 266 | ||
GenBank mRNA | AF143241.1 | - | 2 | 223 | ||
AK048644.1 | - | 3 | 274 | |||
AK185320.1 | - | 1 | 18 | |||
AK192226.1 | - | 1 | 17 | |||
BC042941.1 | - | 3 | 266 | 6493571 |