Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pard6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047435.2 | NP_001040900.1 | 3 | 1114 | ||
NM_001047436.2 | NP_001040901.1 | 3 | 1130 | |||
NM_001286344.1 | NP_001273273.1 | 3 | 1099 | |||
NM_001286345.1 | NP_001273274.1 | 3 | 1027 | |||
NM_019695.3 | NP_062669.2 | 3 | 1117 | |||
GenBank mRNA | AF070970.1 | - | 3 | 1113 | ||
AK003030.1 | - | 3 | 1083 | |||
AK158848.1 | - | 3 | 1112 | |||
BC049593.1 | - | 3 | 1091 | 6771847 | ||
BC061114.1 | - | 3 | 1027 | 6535523 |