Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019718.2 | NP_062692.1 | 4, 5 | 366 | ||
GenBank mRNA | AF143241.1 | - | 3, 4 | 323 | ||
AK048644.1 | - | 4, 5 | 374 | |||
AK192226.1 | - | 2, 3 | 118 | |||
BC042941.1 | - | 4, 5 | 366 | 6493571 |