Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mmp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019724.3 | NP_062698.2 | 6 | 1311 | ||
GenBank mRNA | AB021228.1 | - | 6 | 1126 | ||
AF282844.1 | - | 6 | 1078 | |||
AK048543.1 | - | 6 | 1191 | |||
AK052039.1 | - | 1 | 2683 | |||
AK053598.1 | - | 5 | 3686 | |||
AK077939.1 | - | 1354 | ||||
BC057926.1 | - | 6 | 1313 | 5292478 |