Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Git2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077359.1 | NP_001070827.1 | 2 | 281 | ||
NM_001077360.1 | NP_001070828.1 | 2 | 280 | |||
NM_019834.3 | NP_062808.3 | 2 | 280 | |||
GenBank mRNA | AF148693.1 | - | 2 | 154 | ||
AK005365.1 | - | 2 | 222 | |||
AK017943.1 | - | 2 | 257 | |||
AK148029.1 | - | 2 | 284 | |||
AK220323.1 | - | 2 | 281 | |||
BC043062.1 | - | 2 | 280 | 5705391 | ||
BC056993.1 | - | 2 | 280 | 5705391 |