Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242558.1 | NP_001229487.1 | 2 | 281 | ||
NM_019825.3 | NP_062799.2 | 2 | 281 | |||
GenBank mRNA | AF216186.1 | - | 1 | 172 | ||
AK144338.1 | - | 4 | 520 | |||
AK171864.1 | - | 2 | 442 | |||
BC031113.1 | - | 2 | 285 | |||
BC083187.1 | - | 2 | 281 | 30544739 | ||
JF707634.1 | - | 1 | 140 | |||
JF707635.1 | - | 1 | 140 | |||
JF707636.1 | - | 1 | 140 |