Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177965.1 | NP_001171436.1 | 8 | 762 | ||
NM_019870.3 | NP_063923.1 | 8 | 671 | |||
GenBank mRNA | AJ627039.1 | - | 7 | 490 | ||
AK003806.1 | - | 8 | 699 | |||
AK009697.1 | - | 8 | 673 | |||
AK014469.1 | - | 8 | 600 | |||
AY183136.1 | - | 8 | 640 | |||
AY183137.1 | - | 8 | 731 | |||
BC027219.1 | - | 8 | 663 | 3673739 |