Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019835.2 | NP_062809.2 | 8 | 1118 | ||
GenBank mRNA | AB004786.1 | - | 8 | 1119 | ||
AF142673.1 | - | 8 | 972 | |||
AK151469.1 | - | 5 | 588 | |||
AK152652.1 | - | 5 | 588 |