Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlycd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019966.2 | NP_064350.2 | 5 | 1579 | ||
GenBank mRNA | AJ131522.1 | - | 1 | 442 | ||
AK051564.1 | - | 5 | 1530 | |||
AK082479.1 | - | 5 | 1554 | |||
AK171095.1 | - | 4 | 2413 | |||
BC004764.1 | - | 5 | 1579 | 3155699 | ||
BC092290.1 | - | 2 | 750 |