Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Klk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177373.1 | NP_001170844.1 | 5 | 744 | ||
NM_019974.2 | NP_064358.1 | 4 | 710 | |||
GenBank mRNA | AB016226.1 | - | 5 | 594 | ||
AB016227.1 | - | 5 | 704 | |||
AK009360.1 | - | 5 | 745 | |||
AK009451.1 | - | 1 | 35 | |||
AK009659.1 | - | 5 | 719 | |||
AK009720.1 | - | 5 | 745 | |||
AK019102.1 | - | 1 | 35 | |||
BC115518.2 | - | 5 | 580 | |||
BC148310.2 | - | 4 | 419 |