Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Habp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019986.3 | NP_064370.2 | 2 | 565 | ||
GenBank mRNA | AF227684.1 | - | 2 | 508 | ||
AK016800.1 | - | 2 | 608 | |||
AK144206.1 | - | 2 | 549 | |||
BC082806.1 | - | 2 | 567 | |||
BC119460.2 | - | 1 | 62 | |||
BC148304.2 | - | 1 | 69 |