Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mllt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019914.4 | NP_064298.1 | 2 | 1408 | ||
GenBank mRNA | AB000733.1 | - | 2 | 982 | ||
AK046005.1 | - | 2 | 912 | |||
AK152708.1 | - | 2 | 1408 | |||
AK158495.1 | - | 2 | 1034 | |||
BC022963.1 | - | 2 | 849 | 5365628 | ||
U95498.1 | - | 2 | 970 |