Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019938.3 | NP_064322.1 | 6 | 807 | ||
GenBank mRNA | AB029919.1 | - | 6 | 807 | ||
AK133053.1 | - | 5 | 863 | |||
BC138245.1 | - | 6 | 759 | 8861264 | ||
BC138246.1 | - | 6 | 759 | 8861265 |