Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pate4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020264.4 | NP_064660.2 | 1, 2 | 113 | ||
GenBank mRNA | AF134204.1 | - | 1, 2 | 74 | ||
AK020540.1 | - | 1, 2 | 97 | |||
BC120764.1 | - | 1, 2 | 113 | 40129687 | ||
BC120766.1 | - | 1, 2 | 113 | 40129689 |