Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacstd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020047.3 | NP_064431.2 | 1 | 789 | ||
GenBank mRNA | AK076410.1 | - | 1 | 764 | ||
AK077370.1 | - | 1 | 764 | |||
AK085753.1 | - | 1 | 764 | |||
BC117719.1 | - | 1 | 753 | 40088161 | ||
BC117720.1 | - | 1 | 753 | 40088163 |