Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020050.1 | NP_064434.1 | 5 | 807 | ||
GenBank mRNA | AK007430.1 | - | 5 | 775 | ||
AK009147.1 | - | 5 | 742 | |||
AK049882.1 | - | 5 | 894 | |||
AK077253.1 | - | 5 | 763 | |||
AK147127.1 | - | 5 | 823 | |||
AK207353.1 | - | 1 | 138 | |||
AK215177.1 | - | 1 | 138 | |||
BC002208.1 | - | 5 | 791 | 3490055 |