Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FKBP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017946.3 | NP_060416.1 | 1 | 461 | ||
NR_046478.1 | 1 | 461 | ||||
NR_046479.1 | 1 | 461 | ||||
GenBank mRNA | AK000738.1 | - | 1 | 313 | ||
AK309552.1 | - | 1 | 322 | |||
AY358643.1 | - | 1 | 341 | |||
BC005206.1 | - | 1 | 344 | 4042173 | ||
DA166879.1 | - | 1 | 462 | |||
DB498479.1 | - | 1 | 81 | |||
JQ797475.1 | - | 1 | 168 |