Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GTF2IRD1 SIRNA Assays ›
Gene Symbol:
Gtf2ird1
Gene Name:
general transcription factor II I repeat domain-containing 1
Gene Aliases:
1700012P16Rik, BEN, Cream1, ESTM9, GTF3, Gtf2il, MusTRD1, Tg(Alb1-Myc)166.8Sst, WBSCR11, X83320
Chromosome Location:
Chr.5: 134357661 - 134456716 on Build GRCm38
UniGene ID:
Mm.332735
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
75037
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Gtf2ird1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001081462.2
NP_001074931.1
3
429
NM_001081463.2
NP_001074932.1
3
429
NM_001081464.2
NP_001074933.1
3
429
NM_001081465.1
NP_001074934.1
3
372
NM_001081466.2
NP_001074935.1
3
429
NM_001081467.2
NP_001074936.1
3
429
NM_001081468.2
NP_001074937.1
3
429
NM_001081469.2
NP_001074938.1
3
429
NM_001081470.2
NP_001074939.1
3
429
NM_001244936.1
NP_001231865.1
3
429
NM_020331.3
NP_065064.2
3
429
GenBank mRNA
AF247161.1
-
3
289
AF257475.1
-
3
405
AF260133.1
-
3
400
AF343348.1
-
3
297
AF343349.1
-
3
303
AF343350.1
-
3
299
AF343351.1
-
3
297
AF497637.1
-
3
289
AF497638.1
-
3
289
AF497639.1
-
3
289
AF497640.1
-
3
297
AF497641.1
-
3
303
AF497642.1
-
3
299
AK051310.1
-
3
393
AK078488.1
-
3
297
AK079487.1
-
3
369
AK147196.1
-
3
412
AY030287.1
-
3
400
AY030288.1
-
3
400
AY030289.1
-
3
400
BC050822.1
-
3
424
6393328
BC057091.1
-
3
251
5703300
BC058964.1
-
3
251
5703300
BC083150.1
-
3
331
30459984
CJ048288.1
-
3
372
Back To Top
More Information
Panther Classification:
Molecular Function -
general transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
embryo development
transition between slow and fast fiber
DNA binding
transcription factor activity, sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0