Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020593.2 | NP_065618.1 | 1, 2 | 164 | ||
NM_212433.1 | NP_997598.1 | 1, 2 | 164 | |||
GenBank mRNA | AF233226.1 | - | 1, 2 | 103 | ||
AK004544.2 | - | 1, 2 | 116 | |||
AK009024.1 | - | 1, 2 | 104 | |||
AK019550.1 | - | 1, 2 | 104 | |||
AK028048.1 | - | 1, 2 | 126 | |||
AK031185.1 | - | 1, 2 | 103 | |||
AK031428.1 | - | 1, 2 | 124 | |||
AK049870.1 | - | 1, 2 | 122 | |||
AK050195.1 | - | 1, 2 | 116 | |||
AK085686.1 | - | 1, 2 | 119 | |||
AK150097.1 | - | 1, 2 | 116 | |||
AK151130.1 | - | 1, 2 | 115 | |||
AK151857.1 | - | 1, 2 | 115 | |||
AK152780.1 | - | 1, 2 | 115 | |||
AK167075.1 | - | 1, 2 | 102 | |||
AK215292.1 | - | 1, 2 | 108 | |||
AK215501.1 | - | 1, 2 | 108 | |||
BC010212.1 | - | 1, 2 | 65 | |||
BC050758.1 | - | 1, 2 | 117 | |||
BC058253.1 | - | 1, 2 | 121 | 6465107 | ||
BY105498.1 | - | 1, 2 | 164 |