Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085415.1 | NP_001078884.1 | 3 | 272 | ||
NM_001085416.1 | NP_001078885.1 | 3 | 272 | |||
NM_001085417.1 | NP_001078886.1 | 3 | 625 | |||
NM_020589.2 | NP_065614.2 | 3 | 625 | |||
GenBank mRNA | AB041616.1 | - | 3 | 407 | ||
AB076746.1 | - | 3 | 493 | |||
AK032551.1 | - | 3 | 266 | |||
AK033146.1 | - | 3 | 273 | |||
AK080480.1 | - | 3 | 243 | |||
AK140205.1 | - | 3 | 264 | |||
AK163508.1 | - | 2 | 207 | |||
AK182379.1 | - | 3 | 225 | |||
BC029859.1 | - | 3 | 269 | 4457967 | ||
BC140975.1 | - | 3 | 310 | 9055587 | ||
CK791410.1 | - | 3 | 234 |