Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020593.2 | NP_065618.1 | 2, 3 | 267 | ||
NM_212433.1 | NP_997598.1 | 2, 3 | 267 | |||
GenBank mRNA | AF233226.1 | - | 2, 3 | 206 | ||
AK004544.2 | - | 2, 3 | 219 | |||
AK009024.1 | - | 2, 3 | 206 | |||
AK019550.1 | - | 2, 3 | 207 | |||
AK028048.1 | - | 2, 3 | 229 | |||
AK031185.1 | - | 2, 3 | 206 | |||
AK031428.1 | - | 2, 3 | 227 | |||
AK049870.1 | - | 2, 3 | 225 | |||
AK050195.1 | - | 2, 3 | 219 | |||
AK085686.1 | - | 2, 3 | 222 | |||
AK150097.1 | - | 2, 3 | 219 | |||
AK151130.1 | - | 2, 3 | 218 | |||
AK151857.1 | - | 2, 3 | 218 | |||
AK152780.1 | - | 2, 3 | 218 | |||
AK167075.1 | - | 2, 3 | 205 | |||
AK215501.1 | - | 2, 3 | 211 | |||
BC010212.1 | - | 2, 3 | 168 | |||
BC050758.1 | - | 2, 3 | 220 | |||
BC058253.1 | - | 2, 3 | 224 | 6465107 | ||
BY105498.1 | - | 2, 3 | 268 |