Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFC2-KCTD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001203260.1 | NP_001190189.1 | 4 | 953 | ||
NM_001203261.1 | NP_001190190.1 | 3 | 858 | |||
NM_001203262.1 | NP_001190191.1 | 3 | 809 | |||
GenBank mRNA | BG723290.1 | - | 3 | 506 | ||
FY211285.1 | - | 4 | 507 | |||
FY212410.1 | - | 3 | 363 | |||
FY213449.1 | - | 4 | 507 |
Gene Symbol: KCTD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282406.1 | NP_001269335.1 | 3 | 299 | ||
NM_023930.3 | NP_076419.2 | 2 | 188 | |||
GenBank mRNA | AK313892.1 | - | 3 | 299 | ||
BC001062.2 | - | 2 | 154 | 2819500 | ||
BC001929.1 | - | 2 | 154 | 2819500 | ||
CB993622.1 | - | 2 | 235 |