Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rangrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285441.1 | NP_001272370.1 | 3 | 379 | ||
NM_001285443.1 | NP_001272372.1 | 3 | 475 | |||
NM_021329.3 | NP_067304.1 | 3 | 475 | |||
GenBank mRNA | AF243512.1 | - | 3 | 446 | ||
AK010288.1 | - | 3 | 475 | |||
BB618250.1 | - | 3 | 473 | |||
BC018290.1 | - | 3 | 378 | |||
BC038470.1 | - | 3 | 456 | 5344659 | ||
BC087921.1 | - | 3 | 440 | 6772909 |