Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021335.3 | NP_067310.1 | 3 | 234 | ||
GenBank mRNA | AK005107.1 | - | 3 | 233 | ||
AK011274.1 | - | 3 | 265 | |||
AK012944.1 | - | 243 | ||||
AK013871.1 | - | 3 | 235 | |||
BC026794.1 | - | 3 | 253 | 3483683 |