Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asic5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021370.2 | NP_067345.1 | 3 | 463 | ||
GenBank mRNA | Y19035.1 | - | 3 | 462 |